ITW Dymon do-it ALL Foaming Germicidal Cleaner is an EPA-registered aerosol for cleaning, disinfecting, and deodorizing hard, non-porous surfaces. Its clinging foam formula ensures extended contact time for effective sanitization against a broad spectrum of bacteria and viruses, including SARS-CoV-2.
- Cleans, disinfects, and deodorizes in one step
- Foam clings to vertical surfaces for extended contact time
- EPA registered for proven efficacy
- Controls mold, mildew, and associated odors
- Safe for use in healthcare, institutional, and various environments
- Effective against SARS-CoV-2, HIV-1, Human Coronavirus, Herpes Simplex, Influenza A2, Staphylococcus aureus, Pseudomonas aeruginosa, and Salmonella choleraesuis
- Ideal for bathroom fixtures, tile, shower stalls, and diverse hard surfaces including plastic, leather, and painted finishes
- Contains no abrasives