Learn More
Youngstown Barrel & Drum Plastic 12 gal OH pail White

Description
Store or ship solids.
Specifications
Specifications
| Product Type | Drum |
| Description | Drum |
| Capacity (English) | 12 gal. |
| Color | White |
| For Use With (Application) | For Transport of Hazardous Solids |
| Material | HDPE |
| Includes | Lid |
| Shape | Round |
| Height (English) | 21.44 in. |
| Height (Metric) | 544.58 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.