Learn More
Ubpbio Empty 12ml capacity column for gravity purification, 5 columns per pack
Catalog No. 502339284
Encompass_Preferred
This column serves as a tool for gravity purification using small quantities of resin. The package contains 5 columns with their top and end caps. The PLASTIC 1ML COLUMNs are polypropylene with a total volume of 12 ml and contain a polyethylene frit with a nominal pore size of 20 µm. They are specially designed for working with bed volumes of 0.5-2.0 ml.
Catalog No. 50-233-9284 Supplier Ubpbio Supplier No. P9020
This column serves as a tool for gravity purification using small quantities of resin. The package contains 5 columns with their top and end caps. The PLASTIC 1ML COLUMNs are polypropylene with a total volume of 12 ml and contain a polyethylene frit with a nominal pore size of 20 µm. They are specially designed for working with bed volumes of 0.5-2.0 ml.