Learn More
Ubpbio Empty 35ml capacity column for gravity purification, 5 columns per pack
Catalog No. 502339303
Encompass_Preferred
This column serves as a tool for gravity purification using small quantities of resin. The package contains 5 columns with their top and end caps. The PLASTIC 5ML COLUMN is polypropylene with a total volume of 35 ml and contain a polyethylene frit with a nominal pore size of 20 µm. This column is specially designed for working with bed volumes of 2-6 ml.
Catalog No. 50-233-9303 Supplier Ubpbio Supplier No. P9030
This column serves as a tool for gravity purification using small quantities of resin. The package contains 5 columns with their top and end caps. The PLASTIC 5ML COLUMN is polypropylene with a total volume of 35 ml and contain a polyethylene frit with a nominal pore size of 20 µm. This column is specially designed for working with bed volumes of 2-6 ml.