Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Plastic .6 gal OH pail White

Description
Store or ship smaller quantities of solids.
Specifications
Specifications
| Certifications/Compliance | 1H2/Y4/S |
| Product Type | Open Head |
| Description | Open Head |
| Capacity (English) | 0.6 gal. |
| Color | White |
| For Use With (Application) | PACKAGING FOR HAZARDOUS SOLIDS |
| Material | HDPE |
| Includes | Screw Lid |
| Shape | Round |
| Height (English) | 7.25 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?