Learn More
MSC Dow Corning 99180649 10.1 oz Cartridge Black RTV Silicone Joint Sealant -49 to 392°F Operating Temp, 75 min Tack Free Dry Time, 24 to 72 hr Full Cure Time, Series 739
Catalog No. 501305410
Encompass
Dow Corning 99180649 10.1 oz Cartridge Black RTV Silicone Joint Sealant -49 to 392°F Operating Temp, 75 min Tack Free Dry Time, 24 to 72 hr Full Cure Time, Series 739
Catalog No. 50-130-5410 Supplier MSC Supplier No. 00232009
Dow Corning 99180649 10.1 oz Cartridge Black RTV Silicone Joint Sealant -49 to 392°F Operating Temp, 75 min Tack Free Dry Time, 24 to 72 hr Full Cure Time, Series 739