Learn More
Cole-Parmer Bags Ldpe 10x12 500/pk - Polyethylene (Clear)
Supplier:  Cole-Parmer 0650516
Encompass_Preferred
Constructed of durable LDPE, these general-purpose bags are ideal for a wide range of applications. Seal with portable heat sealer or bag ties. The portable heat sealer is available as part numbers 03014-00 or -02 and the plastic twist ties are available as part number 06831-15.
Catalog No. NC1877452