Learn More
Cole-Parmer Hollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk
Catalog No. NC1118566 Shop All Cole-Parmer Products
Encompass_Preferred
Techne Hollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk, Reduce vapor and heat loss by 75% with one layer of bath ball covers, Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation.
Catalog No. NC1118566 Supplier Cole-Parmer Supplier No. 0682125
Techne Hollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk, Reduce vapor and heat loss by 75% with one layer of bath ball covers, Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation.