Learn More
McMaster-Carr Plastic Barbed Tube Fitting for Food&Beverage
Catalog No. NC2748885
Encompass
Plastic Barbed Tube Fitting for Food&Beverage Polypropylene Reducer for 1/8" x 1/16" Tube ID
Catalog No. NC2748885 Supplier McMaster-Carr Supplier No. 5116K193
Plastic Barbed Tube Fitting for Food&Beverage Polypropylene Reducer for 1/8" x 1/16" Tube ID