Learn More
Lab Storage Systems Inc PLASTIC BLOCK MAILER
SDP
Catalog No. NC0258625
Encompass
Transport container designed specifically for paraffin blocks. Made of PVC 6 block capacity Attached cover snaps securely for transport yet is easy to open and close Transparent container allows for easy viewing of block identification on front & sides Fits all standard size tissue & biopsy cassettes Packaged 50 per case.
Catalog No. NC0258625 Supplier Lab Storage Systems Inc Supplier No. 4776
Transport container designed specifically for paraffin blocks. Made of PVC 6 block capacity Attached cover snaps securely for transport yet is easy to open and close Transparent container allows for easy viewing of block identification on front & sides Fits all standard size tissue & biopsy cassettes Packaged 50 per case.