Learn More
Innovative Med Tech Plastic Blood Gas Capillary - 230 μL, 2.33 x 150 mm, 100 IU/mL
Catalog No. 22564341
Change view
Click to view available options
Capacity (Metric):
230 μL
Catalog No. Capacity (Metric)
22-564-341 230 μL
1 options
Catalog No. 22-564-341 Supplier Innovative Med Tech Supplier No. 102155200
Add to Cart
Add to Cart
Fracture-proof, chemically neutral plastic capillary designed to reduce injury and infection risk during blood gas sample collection.
  • Made from fracture-proof plastic to minimize breakage.
  • Chemically neutral material - compatible with blood gas samples.
  • Reduces risk of injury during blood collection.
  • Lowers infection risk associated with sampling.
  • Ideal for safe blood gas capillary collection.
Specifications
Color Clear
Product Type Capillary Tube
Additive Balanced Li-Heparin
Material PVC
Sterility Non-sterile
Closure Type Cap
Capacity (Metric) 230 μL
Closure Included No