Learn More
Hach Company Acrylic BOD Bottle Stoppers, 25/pk for 300 mL BOD Bottles
Catalog No. NC2065731 Shop All Hach Company Products
Encompass
Unbreakable acrylic stoppers designed for 300 mL BOD bottles. Pack of 25 stoppers provides a reliable seal that prevents oxygen diffusion into or out of samples, reduces cleaning and downtime when used with disposable bottles, and is recyclable.
  • Designed for 300 mL BOD bottles.
  • Pack of 25 stoppers.
  • Unbreakable acrylic construction.
  • Prevents oxygen diffusion in or out of the sample.
  • No cleaning required when used with disposable bottles.
  • Recyclable material.
Catalog No. NC2065731 Supplier Hach Company Supplier No. 2943900
Unbreakable acrylic stoppers designed for 300 mL BOD bottles. Pack of 25 stoppers provides a reliable seal that prevents oxygen diffusion into or out of samples, reduces cleaning and downtime when used with disposable bottles, and is recyclable.
  • Designed for 300 mL BOD bottles.
  • Pack of 25 stoppers.
  • Unbreakable acrylic construction.
  • Prevents oxygen diffusion in or out of the sample.
  • No cleaning required when used with disposable bottles.
  • Recyclable material.