Learn More
Electron Microscopy Sciences Rectangular Plastic Box with Friction-Fitted Lid
SDP
Catalog No. 5033155
Encompass_Preferred
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Friction fitting lid.
Length: 3-3/16"
Width: 6-13/16
Height: 1-5/8"
Catalog No. 50-331-55 Supplier Electron Microscopy Sciences Supplier No. 6431812
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Friction fitting lid.
Length: 3-3/16"
Width: 6-13/16
Height: 1-5/8"