Learn More
Electron Microscopy Sciences Rectangular Plastic Box with Friction-Fitted Lid
SDP
Catalog No. 5033154
Encompass_Preferred
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Friction fitting lid.
Length: 4-1/4"
Width: 1-1/2
Height: 1"
Catalog No. 50-331-54 Supplier Electron Microscopy Sciences Supplier No. 6431814
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Friction fitting lid.
Length: 4-1/4"
Width: 1-1/2
Height: 1"