Learn More
Electron Microscopy Sciences Rectangular Plastic Box with Hinged Lid Snap Lock
SDP
Catalog No. 5027441
Encompass_Preferred
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Hinged lid, snap lock.
Length: 6" x 4"
Width: 4"
Height: 1-1/4"
Lid: 1/4"
Catalog No. 50-274-41 Supplier Electron Microscopy Sciences Supplier No. 64322
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Hinged lid, snap lock.
Length: 6" x 4"
Width: 4"
Height: 1-1/4"
Lid: 1/4"