Learn More
Electron Microscopy Sciences Rectangular Plastic Box with Hinged Lid Snap Lock
SDP
Catalog No. 5027442
Encompass_Preferred
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Hinged lid, snap lock.
Length: 2-7/8"
Width: 2"
Height: 9/16"
Catalog No. 50-274-42 Supplier Electron Microscopy Sciences Supplier No. 64320
Finally – the right shape for all those long and narrow items. Friction fitting lids or hinged-lids with snap locks available. Molded from polystyrene for optimum clarity and rigidity. Hinged lid, snap lock.
Length: 2-7/8"
Width: 2"
Height: 9/16"