Learn More
Electron Microscopy Sciences Plastic Box, Round, Rd2
SDP
Catalog No. 5033158
Encompass_Preferred
Super for storing many items that just don't seem to fit in square or rectangular boxes. Supplied with friction-fitting lid. Especially designed for storing little pieces that constantly are misplaced. Ideal for specimen blocks, apertures etc. Molded from polystyrene for optimum clarity and rigidity. Polystyrene with friction-fitting lids.
Catalog No. 50-331-58 Supplier Electron Microscopy Sciences Supplier No. 64332
Super for storing many items that just don't seem to fit in square or rectangular boxes. Supplied with friction-fitting lid. Especially designed for storing little pieces that constantly are misplaced. Ideal for specimen blocks, apertures etc. Molded from polystyrene for optimum clarity and rigidity. Polystyrene with friction-fitting lids.