Learn More
Grainger Starrett Plastic Case, For S831E Series, Small Hole Gage, 1 ea
Catalog No. 502786629
Encompass_Preferred
This plastic case is designed specifically for housing the S831E series small hole gage set, providing a durable solution for organizing and protecting precision measuring tools.
  • Provides a dedicated storage solution for small hole gage sets
  • Designed for the organization and protection of precision instruments
  • Compatible with S831E series gage sets
Catalog No. 50-278-6629 Supplier Grainger Supplier No. 197V09
This plastic case is designed specifically for housing the S831E series small hole gage set, providing a durable solution for organizing and protecting precision measuring tools.
  • Provides a dedicated storage solution for small hole gage sets
  • Designed for the organization and protection of precision instruments
  • Compatible with S831E series gage sets