Learn More
Grainger Starrett Small Hole Gage Set, 0.125 in. to 0.5 in., 5 Pieces
Catalog No. 502786426
Encompass_Preferred
This small hole gauge set is designed for precise internal measurements in tight spaces. Each of the five gauges features a precision-ground radius that ensures 2-point contact for accurate readings.
  • Includes 830 A, B, C, D, and E gauges
  • Measurement range of 0.125 in. to 0.5 in.
  • Half-ball gauge design with flat bottoms
  • Graduation measurements in inches
  • Comes in a plastic case
Catalog No. 50-278-6426 Supplier Grainger Supplier No. 197V08
This small hole gauge set is designed for precise internal measurements in tight spaces. Each of the five gauges features a precision-ground radius that ensures 2-point contact for accurate readings.
  • Includes 830 A, B, C, D, and E gauges
  • Measurement range of 0.125 in. to 0.5 in.
  • Half-ball gauge design with flat bottoms
  • Graduation measurements in inches
  • Comes in a plastic case