Learn More
Lovibond™ Plastic Cell
Catalog No. 09053294 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-053-294 10 mL
1 options
Catalog No. 09-053-294 Supplier Lovibond™ Supplier No. 145505
Add to Cart
Add to Cart
The set includes five 10 mL units with a 13.5 mm path length and grey lids.
  • Used with Lovibond CHECKIT™ and Comparator 2000 test kits
  • Refer to the instruction manual for your color disc to ensure this is the correct path length
Specifications
pH Range 0 to 14
Volume (Metric) 10 mL
Type Plastic Cell