Learn More
MSC Novus Plastic Polish, No. 1, Clean and Shine, 8 oz
Catalog No. 502868931
Encompass
Novus Plastic Polish #1 is an organic liquid/water mixture designed to clean and restore plastic surfaces. This polisher effectively cleans and shines plastics, making it suitable for various applications including the restoration of acrylics and cleaning 3D printer trays.
  • Cleans and restores plastic surfaces
  • Causes skin and eye irritation
  • Provided in an 8 oz bottle
Catalog No. 50-286-8931 Supplier MSC Supplier No. 74766544
Novus Plastic Polish #1 is an organic liquid/water mixture designed to clean and restore plastic surfaces. This polisher effectively cleans and shines plastics, making it suitable for various applications including the restoration of acrylics and cleaning 3D printer trays.
  • Cleans and restores plastic surfaces
  • Causes skin and eye irritation
  • Provided in an 8 oz bottle