Learn More
Research Products International Corp Plastic Cover for Aluminum Block which holds 40 x 0.5ml or 15 1.5-2.0ml Micro-tubes
SDP
Catalog No. 501977970
Encompass
Optional plastic cover for aluminum blocks which hold 40 x 0.5ml or 15 1.5-2.0ml Micro-tubes. For use with chilling/heating dry baths.
Catalog No. 50-197-7970 Supplier Research Products International Corp Supplier No. 248814
Optional plastic cover for aluminum blocks which hold 40 x 0.5ml or 15 1.5-2.0ml Micro-tubes. For use with chilling/heating dry baths.