Learn More
Ihc World LLC PLASTIC COVERSLIP 24X60 100/PK
SDP
Catalog No. NC0052976
Encompass
Used for protein digestion, antigen retrieval protocols, antibody incubation and other staining procedures where require minimum reagent usage. Temperature range: 0 to 65 C. Size: pack of 100pcs.
Catalog No. NC0052976 Supplier Ihc World LLC Supplier No. IW2601
Used for protein digestion, antigen retrieval protocols, antibody incubation and other staining procedures where require minimum reagent usage. Temperature range: 0 to 65 C. Size: pack of 100pcs.