Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Plastic Disabled Access Sign: ACCESSIBLE

Catalog No. 19109478 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
6 x 9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-109-478 6 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-109-478 Supplier Brady™ Supplier No. 103861

missing translation for 'validMainframeMsg'

Improve workplace safety with proper signage and markings. This Brady™ mounted sign is a durable, clear and effective tool to mark disabled access ways.

  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information

Specifications

Material Plastic
Quantity 1 Ea.
Height (English) 9 in.
Width (English) 6 in.
Shape Rectangular
Dimensions (L x W) 6 x 9 in.
Displayed Text Handicap Accessible
Color White on Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.