Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Plastic Disposable Generator Probe

Catalog No. 15341006 Shop All Omni International Inc Products
Change view
Click to view available options
Packaging Quantity:
100 Pk.
25 Pk.
50 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
15-341-006 100 Pk.
15-341-004 25 Pk.
15-341-002 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-341-006 Supplier Fisherbrand™ Supplier No. 34550F
Add to Cart
Add to Cart

Significantly reduces the hassle of disassembly and maintenance of traditional stainless-steel generators

  • Cost-effective solution
  • <1% of the cost of stainless steel probes
  • Significantly reduces cross contamination and associated risks
  • Popular for use with low-analyte detection assays, such as PCR
  • Commonly used for RNA and DNA extraction
  • Simple 2-piece construction
  • Compatible with Fisherbrand 150 and Fisherbrand 850 Homogenizers

Specifications

Product Type Probe
Capacity (Metric) 10 to 100 mg
Diameter (Metric) Probe 5 mm
Dimensions (L x W x H) 5 x 7 mm
Autoclavable Autoclavable
Blade Material Polypropylene
Material Polycarbonate, Polyetherimide
Bottom Type Flat
Closure Color Clear
Color Blue
Packaging Quantity 100 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.