Learn More
Bellco Glass, Inc. Plastic Dome Cover Hot Shaker
SDP
Supplier:  Bellco Glass, Inc. 774612400
Encompass_Preferred
Clear Plexiglass dome shape cover for Single Pan Hot Shakers. Can withstand continuous heat (180°-200°F) for long periods of time. Dimensions: 21 5/8" wide x 13" deep x 4 3/4" height With 1/2" hole for Thermowell Material: 3/16″ Thick Clear Plexiglass . MADE IN THE USA
Catalog No. 50-194-6170