Learn More
Grainger Plastic Door Handle 80/20 10 /35 Series
Catalog No. 502764037
Encompass_Preferred
The 80/20 10-35 Series Plastic Door Handle is a black plastic door handle made from acetal copolymer plastic. It has a curved design with a return to provide a comfortable grip. The handle measures 4" long x 1-3/4" wide and projects 2-1/4" from the door surface. It has a 1/4" diameter mounting screw on a 2-1/2" center to center. The handle is rated for interior use on doors up to 1-3/8" thick and has a temperature range of -20°F to 180°F. This durable handle provides an economical option for industrial doors, cabinets and enclosures.
Catalog No. 50-276-4037 Supplier Grainger Supplier No. 16U361
The 80/20 10-35 Series Plastic Door Handle is a black plastic door handle made from acetal copolymer plastic. It has a curved design with a return to provide a comfortable grip. The handle measures 4" long x 1-3/4" wide and projects 2-1/4" from the door surface. It has a 1/4" diameter mounting screw on a 2-1/2" center to center. The handle is rated for interior use on doors up to 1-3/8" thick and has a temperature range of -20°F to 180°F. This durable handle provides an economical option for industrial doors, cabinets and enclosures.