Learn More
MSC Quantum Storage Systems Drawer Cabinet, 60 Drawers, Polypropylene, 1 ea
Catalog No. 5000514663
Encompass
This 60-drawer cabinet system provides efficient organization and storage for small parts. Engineered with a heavy-duty, reinforced polypropylene frame, this unit is designed for durability in demanding environments. The clear, plastic drawers allow for quick visual identification of contents and feature textured pulls for easy access.
  • Sixty clear plastic drawers for high-capacity organization
  • Heavy-duty, reinforced polypropylene construction
  • Integrated drawer stops to prevent accidental spills
  • Includes drawer dividers for every drawer to maximize storage versatility
  • Stable design suitable for wall mounting
  • Overall dimensions: 15.88 cm x 38.1 cm x 47.62 cm (6-1/4 in. x 15 in. x 18-3/4 in.)
Catalog No. 50-005-14663 Supplier MSC Supplier No. 41349820
This 60-drawer cabinet system provides efficient organization and storage for small parts. Engineered with a heavy-duty, reinforced polypropylene frame, this unit is designed for durability in demanding environments. The clear, plastic drawers allow for quick visual identification of contents and feature textured pulls for easy access.
  • Sixty clear plastic drawers for high-capacity organization
  • Heavy-duty, reinforced polypropylene construction
  • Integrated drawer stops to prevent accidental spills
  • Includes drawer dividers for every drawer to maximize storage versatility
  • Stable design suitable for wall mounting
  • Overall dimensions: 15.88 cm x 38.1 cm x 47.62 cm (6-1/4 in. x 15 in. x 18-3/4 in.)