Learn More
Uline Plastic Drum Pumps
Catalog No. NC0857648
Encompass
Plastic drum pump with polypropylene body, Viton seals, 2 inch bung adapter, maximum 30 gallon per minute flow rate enables convenient, mess-free transfer of corrosive chemicals, oils, solvents and other industrial liquids from standard 55 gallon drums.
Catalog No. NC0857648 Supplier Uline Supplier No. H1553
Plastic drum pump with polypropylene body, Viton seals, 2 inch bung adapter, maximum 30 gallon per minute flow rate enables convenient, mess-free transfer of corrosive chemicals, oils, solvents and other industrial liquids from standard 55 gallon drums.