Learn More
Biodesign Inc of Ny Filtration Unit, Type B, Three complete units
Catalog No. NC2684803
Encompass
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.
Catalog No. NC2684803 Supplier Biodesign Inc of Ny Supplier No. F109
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.