Learn More
Biodesign Inc of Ny Filtration Unit for 47 mm circles, Type B, One unit, non-toxic plastic that can be repeatedly autoclaved
Catalog No. NC2684800
Encompass
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.
Catalog No. NC2684800 Supplier Biodesign Inc of Ny Supplier No. F107
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.