Learn More
Cole-Parmer Indenti-Plug L800-B Plastic Foam Stopper, 14-20mm Openings, 1000/pk
Catalog No. NC0471237 Shop All Cole-Parmer Products
Encompass_Preferred
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
Catalog No. NC0471237 Supplier Cole-Parmer Supplier No. 0629872
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.