Learn More
MSC Rubbermaid Square Storage Container, 18 QT, Polycarbonate, Clear
Catalog No. 502839287
Encompass
This square food storage container is constructed from durable, clear polycarbonate, offering an 18-quart capacity. Designed for efficiency and hygiene, it features graduated measurements for precise portioning, is dishwasher-safe, impact-resistant, and stackable. Its design aids in HACCP compliance and meets NSF 2 and NSF 3-A standards.
  • Made from dishwasher-safe, impact-resistant polycarbonate
  • Features easy-to-read, color-coded graduated measurements for accurate portioning
  • Square shape provides 25% more storage capacity compared to round containers
  • Designed to assist with HACCP guidelines compliance
  • NSF 2 and NSF 3-A certified
  • BPA free construction
  • Includes durable handles for easy transport
  • Equipped with drain holes for proper washing and drying
  • Stackable for space-saving storage
  • Withstands temperatures from -40°F to 212°F
Catalog No. 50-283-9287 Supplier MSC Supplier No. 58891581
This square food storage container is constructed from durable, clear polycarbonate, offering an 18-quart capacity. Designed for efficiency and hygiene, it features graduated measurements for precise portioning, is dishwasher-safe, impact-resistant, and stackable. Its design aids in HACCP compliance and meets NSF 2 and NSF 3-A standards.
  • Made from dishwasher-safe, impact-resistant polycarbonate
  • Features easy-to-read, color-coded graduated measurements for accurate portioning
  • Square shape provides 25% more storage capacity compared to round containers
  • Designed to assist with HACCP guidelines compliance
  • NSF 2 and NSF 3-A certified
  • BPA free construction
  • Includes durable handles for easy transport
  • Equipped with drain holes for proper washing and drying
  • Stackable for space-saving storage
  • Withstands temperatures from -40°F to 212°F