Learn More
Electron Microscopy Sciences Plastic Forceps
SDP
Catalog No. 5027878
Encompass_Preferred
5" (12.7cm) long, molded, strong, durable, autoclavable up to 275 degree F (135 degree C), serrated tip.
Catalog No. 50-278-78 Supplier Electron Microscopy Sciences Supplier No. 72970
5" (12.7cm) long, molded, strong, durable, autoclavable up to 275 degree F (135 degree C), serrated tip.