Learn More
Fine Science Tools Plastic Forceps, Acetyl, Straight, 11.5 cm
SDP
Catalog No. NC1643876
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.4mm/ Alloy / Material: Acetyl/ Length: 11.5cm
This item is not returnable. View return policy
Catalog No. NC1643876 Supplier Fine Science Tools Supplier No. 1170003
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.4mm/ Alloy / Material: Acetyl/ Length: 11.5cm