Learn More
Fine Science Tools Plastic Forceps, Acetyl, Curved, 11.5 cm
SDP
Catalog No. NC9622764
Encompass
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.4mm/ Alloy / Material: Acetyl/ Length: 11.5cm
Catalog No. NC9622764 Supplier Fine Science Tools Supplier No. 1170004
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.4mm/ Alloy / Material: Acetyl/ Length: 11.5cm