Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Plastic Funnel with Handle, White

Catalog No. 09051493 Shop All Tintometer Group Products
Change view
Click to view available options
Capacity (Metric):
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-051-493 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-051-493 Supplier Lovibond™ Supplier No. 471007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General purpose funnel fits Lovibond sample vials.

  • For methods that require powder to be added with measuring spoons to prevent reagent from being spilled.

Specifications

Capacity (Metric) 10 g
Material Plastic
Height (Metric) 2 in.
Product Type Filling Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.