Learn More
Medex Supply PLASTIC GONIOMETER 12IN
SDP
Supplier:  Medex Supply FAB121000
Encompass_Preferred
•Clear plastic construction permits observation of joint axis and range of motion. •360° head has three scales calibrated to be used with the ISOM system. •One arm has a linear scale with inches and centimeters. •Reads in 1" increments.
Catalog No. NC0074219