Learn More
Grainger 24 to 250mL Plastic Graduated Cylinder, Clear, Height: 315 mm / 12.4 in, 1 EA
Catalog No. NC1816712
Encompass_Preferred
This graduated cylinder helps provide precise measurement of liquids or objects. PMP (polymethylpentene) material offers glasslike clarity, excellent chemical resistance, and heat tolerance.
Catalog No. NC1816712 Supplier Grainger Supplier No. 8ADT5
This graduated cylinder helps provide precise measurement of liquids or objects. PMP (polymethylpentene) material offers glasslike clarity, excellent chemical resistance, and heat tolerance.