Learn More
Fisherbrand™ Plastic Handled Cotton Swabs and Applicators
Catalog No. 22363163 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1/envelope; 1000/cs
1000/pk
2/envelope; 2000/cs
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sterility
22-363-163 2/envelope; 2000/cs Sterile
22-363-162 1/envelope; 1000/cs Sterile
22-363-161 1000/pk Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-363-163
Add to Cart
Add to Cart
This item is not returnable. View return policy
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instrument
  • Sterile- Not suitable for PCR DNA/ RNA testing
  • Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
  • Sterile and non-sterile products available (see product specifications)
  • Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
  • Not suitable for PCR DNA/ RNA testing
Specifications
Product Type Cotton Tip Applicator
Tip Material Cotton
Handle Material Plastic
Sterility Sterile
Quantity 2/envelope; 2000/cs
Length (Metric) 15 cm
Length (English) 6 in.
For Use With (Application) Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments