Learn More
Electron Microscopy Sciences Plastic Hinged Box with 12 Compartments
SDP
Supplier:  Electron Microscopy Sciences 64341
Encompass_Preferred
Organize singular or groups of items in individual compartments. Stored items stay separately and are readily visible at all times. Extra-thick wall construction permits containment of heavy objects; protects delicate items.
Dimensions: 7 x 3-3/4 x 1-1/4"
Compartments: 1-11/16 x 1-1/16 x 1-1/8"
Catalog No. 50-331-40