Learn More
Eldon James Plastic Hose Barb Coupler
Description
- Our medical and pharmaceutical grade plastic tube couplers are RoHS (Restriction of Hazardous Substances) compliant, meet USP Class VI requirements, and are animal derivative-free.
- Transportation and automotive barb couplings meet automotive standards and approvals.
- Eldon James specializes in producing safe, efficient, and high quality products, all of which are made from Eco-friendly resins that will not leach pollutants or contaminate water, liquBarbs, gases, soil, or even other recyclable materials.
Specifications
Specifications
| Description | Class 7 cleanroom manufactured |
| Material | Kynar PVDF |
| Hose Barb | 12 Barb (3/4 in.) |
| Color | Natural |
| Product Type | Straight Coupler |