Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eldon James Plastic Hose Barb Coupler
SDP

Catalog No. 01000833
Change view
Click to view available options
Product Type:
Reduction Coupler
Straight Coupler
Hose Barb:
10 Barb (5/8 in.) to 8 Barb (1/2 in.)
12 Barb (3/4 in.) to 10 Barb (5/8 in.)
12 Barb (3/4 in.) to 6 Barb (3/8 in.)
12 Barb (3/4 in.) to 8 Barb (1/2 in.)
16 Barb (1 in.) to 10 Barb (5/8 in.)
3 Barb (3/16 in.)
6 Barb (3/8 in.)
6 Barb (3/8 in.) to 4 Barb (1/4 in.)
8 Barb (1/2 in.) to 4 Barb (1/4 in.)
8 Barb (1/2 in.) to 6 Barb (3/8 in.)
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Hose Barb
01-000-833 Straight Coupler 3 Barb (3/16 in.)
01-000-824 Reduction Coupler 10 Barb (5/8 in.) to 8 Barb (1/2 in.)
01-000-825 Reduction Coupler 12 Barb (3/4 in.) to 10 Barb (5/8 in.)
01-000-826 Reduction Coupler 12 Barb (3/4 in.) to 6 Barb (3/8 in.)
01-000-827 Reduction Coupler 12 Barb (3/4 in.) to 8 Barb (1/2 in.)
01-000-828 Reduction Coupler 16 Barb (1 in.) to 10 Barb (5/8 in.)
01-000-829 Reduction Coupler 6 Barb (3/8 in.) to 4 Barb (1/4 in.)
01-000-830 Reduction Coupler 8 Barb (1/2 in.) to 4 Barb (1/4 in.)
01-000-831 Reduction Coupler 8 Barb (1/2 in.) to 6 Barb (3/8 in.)
01-000-834 Straight Coupler 6 Barb (3/8 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 01-000-833 Supplier Eldon James Supplier No. C03NK7QC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eldon James features straight couplers and reduction couplers that are perfect for conjoining tubes of any kind. Safe for use in medical practices, our straight couplers and reduction couplers feature innovative single barb technology to stay firmly entrenched in the tubing.

  • Our medical and pharmaceutical grade plastic tube couplers are RoHS (Restriction of Hazardous Substances) compliant, meet USP Class VI requirements, and are animal derivative-free.
  • Transportation and automotive barb couplings meet automotive standards and approvals.
  • Eldon James specializes in producing safe, efficient, and high quality products, all of which are made from Eco-friendly resins that will not leach pollutants or contaminate water, liquBarbs, gases, soil, or even other recyclable materials.

Specifications

Description Class 7 cleanroom manufactured
Material Kynar PVDF
Hose Barb 3 Barb (3/16 in.)
Color Natural
Product Type Straight Coupler
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.