Learn More
Fine Science Tools Instrument Cases, Green, 17 x 8 x 2 cm
SDP
Catalog No. NC1576639
Encompass
Product: Instrument Cases - 17 x 8 x 2cm/ Dimensions: 17 x 8 x 2cm/ Alloy / Material: Plastic/ Color: Green/ Autoclave Safe: Yes/ Unit: Each
Catalog No. NC1576639 Supplier Fine Science Tools Supplier No. 2083001
Product: Instrument Cases - 17 x 8 x 2cm/ Dimensions: 17 x 8 x 2cm/ Alloy / Material: Plastic/ Color: Green/ Autoclave Safe: Yes/ Unit: Each