Learn More
TITAN MEDICAL INSTRUMENTS LLC Plastic Instrument Sterilization Tray Long Medium 4.0" W x 7.5" L x 0.75" H
Catalog No. NC3937210
Encompass
PST Plastic Instrument Sterilization Tray Long Medium Single LevelDesigned to store and protect eye surgery instruments during sterilization. Made in USA. ● Size: 4.0" (102mm) W x 7.5" (190mm) L x 0.75" (19mm) H● All sterilization methods: steam, autoclave, dry heat, flash, EtO, gas plasma, gamma radiation, hydrogen peroxide, cold sterilants, disinfectants, germicides● The unique grid system in all bases, trays, and lids● Stackable trays for easy handling and storage● This is a complete set: base, lid, and silicone mat● Reusable, Non-Sterile● Material: General Electric's ULTEM® resin and surgical grade silicone
Catalog No. NC3937210 Supplier TITAN MEDICAL INSTRUMENTS LLC Supplier No. 4700A
PST Plastic Instrument Sterilization Tray Long Medium Single LevelDesigned to store and protect eye surgery instruments during sterilization. Made in USA. ● Size: 4.0" (102mm) W x 7.5" (190mm) L x 0.75" (19mm) H● All sterilization methods: steam, autoclave, dry heat, flash, EtO, gas plasma, gamma radiation, hydrogen peroxide, cold sterilants, disinfectants, germicides● The unique grid system in all bases, trays, and lids● Stackable trays for easy handling and storage● This is a complete set: base, lid, and silicone mat● Reusable, Non-Sterile● Material: General Electric's ULTEM® resin and surgical grade silicone