Learn More
AirSea USA 5L UN HDPE Plastic Blue Jerrican, 3H1/Y1.9/150. UN Approved Single Packaging Plastic Jerrican– for the transportation of (Liquids) Dangerous Goods. It is also suitable as an inner packaging
Catalog No. NC2101153
Encompass_Preferred
5L UN HDPE Plastic Blue Jerrican, 3H1/Y1.9/150. UN Approved Single Packaging Plastic Jerrican– for the transportation of (Liquids) Dangerous Goods. It is also suitable as an inner packaging. This 5L HDPE UN plastic jerrican (blue) is approved for shipping hazardous goods as liquids within Packing Groups II & III.
This item is not returnable. View return policy
Catalog No. NC2101153 Supplier AirSea USA Supplier No. 22
5L UN HDPE Plastic Blue Jerrican, 3H1/Y1.9/150. UN Approved Single Packaging Plastic Jerrican– for the transportation of (Liquids) Dangerous Goods. It is also suitable as an inner packaging. This 5L HDPE UN plastic jerrican (blue) is approved for shipping hazardous goods as liquids within Packing Groups II & III.