Learn More
Andwin Scientific PLASTIC LARGE BOAT 500/PK
SDP
Catalog No. 50727590
Encompass_Preferred
The large "Anti-Static" Plastic Weighing Boats are ideal for weighing liquid or solid samples. The corners are recessed, which allow users to carefully monitor the pouring of liquids. The weighing boats feature a square shape and a contour bottom. This product is sold in a pack of 500.
Catalog No. 50-727-590 Supplier Andwin Scientific Supplier No. 70042
The large "Anti-Static" Plastic Weighing Boats are ideal for weighing liquid or solid samples. The corners are recessed, which allow users to carefully monitor the pouring of liquids. The weighing boats feature a square shape and a contour bottom. This product is sold in a pack of 500.