Learn More
Elma™ Plastic Lid for Elmasonic S50R Ultrasonic Sieve Cleaner
Description
- Protects against evaporation and splashes
- Serves as noise control barriers and drip trays for wet baskets
- Keeps content free of contaminants
Specifications
Specifications
| Product Type | Lid |
| For Use With (Equipment) | Ultrasonic Cleaner, Elmasonic S50R |
| For Use With (Application) | Protects Against Evaporation and Splashes |
| Material | Plastic |