Learn More
Electron Microscopy Sciences Hard Plastic Microscope Slide Tray
SDP
Catalog No. 50366073
Encompass_Preferred
This durable, ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1"). Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable.
Catalog No. 50-366-073 Supplier Electron Microscopy Sciences Supplier No. 7150310
This durable, ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1"). Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable.