Learn More
MSC Vestil Pail, High-Density Polyethylene, 1 Gallon, Open Head, Plastic Handle
Catalog No. 502839939
Encompass
This open head pail is constructed from 100% prime virgin high-density polyethylene, designed for robust performance in various applications. It offers strength for shipping and durability for demanding storage, making it suitable for both industrial and laboratory environments.
  • Complies with FDA regulations for food contact.
  • Compact and nestable design for simple storage.
  • Features one-piece molded construction with a taper.
  • Suitable for liquids up to 190 degrees Fahrenheit.
  • Ideal for freezer applications.
Catalog No. 50-283-9939 Supplier MSC Supplier No. 91749770
This open head pail is constructed from 100% prime virgin high-density polyethylene, designed for robust performance in various applications. It offers strength for shipping and durability for demanding storage, making it suitable for both industrial and laboratory environments.
  • Complies with FDA regulations for food contact.
  • Compact and nestable design for simple storage.
  • Features one-piece molded construction with a taper.
  • Suitable for liquids up to 190 degrees Fahrenheit.
  • Ideal for freezer applications.